ENA448968390 Enamine

4-chloro-2-(piperidin-1-yl)pyridine

  • MDL number MFCD10703543
  • Cas Number : 1086376-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211672
Availability details for
Quantity
  • Availability
  • Add to Cart