ENA448968405 Enamine

2,2-dichloroethan-1-amine hydrochloride

  • MDL number MFCD27995647
  • Cas Number : 5960-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211812
Availability details for
Quantity
  • Availability
  • Add to Cart