ENA448968407 Enamine

6,6-difluoro-1,4-oxazepane hydrochloride

  • MDL number MFCD21362353
  • Cas Number : 1341039-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211815

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211815
Availability details for
Quantity
  • Availability
  • Add to Cart