ENA448968409 Enamine

methyl(1-phenylpent-4-yn-2-yl)amine

  • MDL number MFCD16698415
  • Cas Number : 1248389-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211817
Availability details for
Quantity
  • Availability
  • Add to Cart