ENA448968410 Enamine

ethyl 5-chloro-3,3-dimethylpentanoate

  • MDL number MFCD24849951
  • Cas Number : 74580-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211819

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211819
Availability details for
Quantity
  • Availability
  • Add to Cart