ENA448968411 Enamine

2-[4-(propan-2-yl)phenoxy]propan-1-amine

  • MDL number MFCD09906790
  • Cas Number : 951915-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211821
Availability details for
Quantity
  • Availability
  • Add to Cart