ENA448968414 Enamine

(piperidin-2-yl)methyl N-propylcarbamate hydrochloride

  • MDL number MFCD28505843
  • Cas Number : 1803562-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211832
Availability details for
Quantity
  • Availability
  • Add to Cart