ENA448968416 Enamine

3-[5-amino-3-(3,4-dimethylphenyl)-1H-pyrazol-1-yl]-1lambda6-thiolane-1,1-dione

  • MDL number MFCD13660430
  • Cas Number : 1152684-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211834
Availability details for
Quantity
  • Availability
  • Add to Cart