ENA448968417 Enamine

2-bromo-1-(4-methylpyridin-2-yl)ethan-1-one

  • MDL number MFCD13173227
  • Cas Number : 759442-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211836

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211836
Availability details for
Quantity
  • Availability
  • Add to Cart