ENA448968424 Enamine

2-ethylcyclobutan-1-one

  • MDL number MFCD00049150
  • Cas Number : 10374-14-8
  • Molecular Weight : 98.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-211844

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211844
Availability details for
Quantity
  • Availability
  • Add to Cart