ENA448968426 Enamine

N-hydroxypyridine-4-carbonimidoyl chloride

  • Cas Number : 4185-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-175405

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-175405
Availability details for
Quantity
  • Availability
  • Add to Cart