ENA448968429 Enamine

2-chloro-4,6-diethylpyrimidine

  • MDL number MFCD22397279
  • Cas Number : 65267-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211855
Availability details for
Quantity
  • Availability
  • Add to Cart