ENA448968430 Enamine

2'-methyl-1,3'-bipyrrolidine

  • MDL number MFCD28505847
  • Cas Number : 1803599-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211857
Availability details for
Quantity
  • Availability
  • Add to Cart