ENA448968441 Enamine

(3-fluoropiperidin-3-yl)methanol hydrochloride

  • MDL number MFCD22690368
  • Cas Number : 1416440-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211889
Availability details for
Quantity
  • Availability
  • Add to Cart