ENA448968443 Enamine

dec-9-en-1-amine hydrochloride

  • MDL number MFCD28505850
  • Cas Number : 1451042-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211891

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211891
Availability details for
Quantity
  • Availability
  • Add to Cart