ENA448968449 Enamine

3-amino-2-methyl-2-phenylpropan-1-ol

  • MDL number MFCD09036472
  • Cas Number : 98459-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211903

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211903
Availability details for
Quantity
  • Availability
  • Add to Cart