ENA448968464 Enamine

2-{[2-(4-aminophenyl)ethyl]amino}ethan-1-ol

  • MDL number MFCD19306115
  • Cas Number : 1225351-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211970
Availability details for
Quantity
  • Availability
  • Add to Cart