ENA448968472 Enamine

1-ethenyl-3,5-dimethoxybenzene

  • MDL number MFCD20384494
  • Cas Number : 40243-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211983

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211983
Availability details for
Quantity
  • Availability
  • Add to Cart