ENA448968479 Enamine

5-bromo-7-chloro-1,3-benzoxazole

  • MDL number MFCD16468585
  • Cas Number : 1226072-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211990
Availability details for
Quantity
  • Availability
  • Add to Cart