ENA448968481 Enamine

4-(propylsulfamoyl)benzene-1-sulfonyl chloride

  • MDL number MFCD09810487
  • Cas Number : 1016532-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211992
Availability details for
Quantity
  • Availability
  • Add to Cart