ENA448968486 Enamine

1-(chloromethyl)cyclopentane-1-carbonitrile

  • MDL number MFCD19232536
  • Cas Number : 112906-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212008
Availability details for
Quantity
  • Availability
  • Add to Cart