ENA448968499 Enamine

sodium 4-bromobenzene-1-sulfinate

  • MDL number MFCD09065037
  • Cas Number : 34176-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181864
Availability details for
Quantity
  • Availability
  • Add to Cart