ENA448968502 Enamine

tricyclo[8.2.1.0,3,8]trideca-3(8),4,6-trien-13-amine

  • MDL number MFCD20700375
  • Cas Number : 1379292-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212055

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212055
Availability details for
Quantity
  • Availability
  • Add to Cart