ENA448968506 Enamine

3-(4-aminopiperidin-1-yl)-1-methylpyrrolidin-2-one dihydrochloride

  • MDL number MFCD28505862
  • Cas Number : 1803562-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212098

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212098
Availability details for
Quantity
  • Availability
  • Add to Cart