ENA448968511 Enamine

2-chloro-1-ethoxy-3-fluorobenzene

  • MDL number MFCD24849963
  • Cas Number : 909122-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212105
Availability details for
Quantity
  • Availability
  • Add to Cart