ENA448968512 Enamine

1,2,4-oxadiazole-3-carbonitrile

  • MDL number MFCD26938432
  • Cas Number : 39512-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212106

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212106
Availability details for
Quantity
  • Availability
  • Add to Cart