ENA448968515 Enamine

2-chloropyridine-3,5-dicarbonitrile

  • MDL number MFCD18802940
  • Cas Number : 172208-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212110
Availability details for
Quantity
  • Availability
  • Add to Cart