ENA448968527 Enamine

6-(2-methylbutan-2-yl)-4,5,6,7-tetrahydro-1,3-benzothiazol-2-amine

  • MDL number MFCD03424584
  • Cas Number : 676348-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212139
Availability details for
Quantity
  • Availability
  • Add to Cart