ENA448968528 Enamine

4-methoxy-2,3,6-trimethylaniline

  • MDL number MFCD18824956
  • Cas Number : 274907-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212141

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212141
Availability details for
Quantity
  • Availability
  • Add to Cart