ENA448968530 Enamine

6-fluoro-3-hydroxypyridine-2-carbonitrile

  • MDL number MFCD18816440
  • Cas Number : 727736-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212143
Availability details for
Quantity
  • Availability
  • Add to Cart