ENA448968534 Enamine

3-methyl-3-[3-(trifluoromethyl)phenyl]pyrrolidine hydrochloride

  • MDL number MFCD27954427
  • Cas Number : 1781945-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212147
Availability details for
Quantity
  • Availability
  • Add to Cart