ENA448968543 Enamine

3-(pyrrolidine-1-carbonyl)pyrrolidine hydrochloride

  • MDL number MFCD20487821
  • Cas Number : 1315592-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212177

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212177
Availability details for
Quantity
  • Availability
  • Add to Cart