ENA448968552 Enamine

4-bromo-3-fluoro-N,N-dimethylbenzamide

  • MDL number MFCD16084889
  • Cas Number : 893420-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212196

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212196
Availability details for
Quantity
  • Availability
  • Add to Cart