ENA448968555 Enamine

1-(1-phenyl-1H-imidazol-2-yl)ethan-1-amine hydrochloride

  • MDL number MFCD28505875
  • Cas Number : 1803604-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212201
Availability details for
Quantity
  • Availability
  • Add to Cart