ENA448968556 Enamine

1-(1-phenyl-1H-imidazol-2-yl)ethan-1-amine

  • MDL number MFCD28505876
  • Cas Number : 1486790-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212202

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212202
Availability details for
Quantity
  • Availability
  • Add to Cart