ENA448968560 Enamine

1-(ethenesulfonyl)-2-methoxyethane

  • MDL number MFCD17255959

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212238
Availability details for
Quantity
  • Availability
  • Add to Cart