ENA448968574 Enamine

4-(trifluoromethyl)cyclohexane-1-sulfonyl chloride

  • MDL number MFCD20410298
  • Cas Number : 1343053-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212274
Availability details for
Quantity
  • Availability
  • Add to Cart