ENA448968589 Enamine

1H,2H,3H,3aH,4H,9H-pyrrolo[2,1-b]quinazolin-9-one

  • MDL number MFCD01082695
  • Cas Number : 22458-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212325

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212325
Availability details for
Quantity
  • Availability
  • Add to Cart