ENA448968595 Enamine

methyl 2-[(piperidin-4-yl)methyl]benzoate hydrochloride

  • MDL number MFCD08460883
  • Cas Number : 1187172-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212342
Availability details for
Quantity
  • Availability
  • Add to Cart