ENA448968597 Enamine

5-methoxy-4,4-dimethylpentan-1-amine

  • MDL number MFCD19374448
  • Cas Number : 1267419-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212345

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212345
Availability details for
Quantity
  • Availability
  • Add to Cart