ENA448968599 Enamine

4-(isoquinolin-1-yl)-1,3-thiazol-2-amine

  • MDL number MFCD26036866
  • Cas Number : 651042-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212357
Availability details for
Quantity
  • Availability
  • Add to Cart