ENA448968604 Enamine

methyl[(4-methyl-4H-1,2,4-triazol-3-yl)methyl]amine

  • MDL number MFCD08059663
  • Cas Number : 936940-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212365

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212365
Availability details for
Quantity
  • Availability
  • Add to Cart