ENA448968613 Enamine

3-(aminomethyl)piperidine-1-carboxamide hydrochloride

  • MDL number MFCD28505891
  • Cas Number : 1803580-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212381
Availability details for
Quantity
  • Availability
  • Add to Cart