ENA448968620 Enamine

5-bromo-3-(2,2,2-trifluoroethyl)-3,4-dihydropyrimidin-4-one

  • MDL number MFCD21746741
  • Cas Number : 1512913-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212393

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212393
Availability details for
Quantity
  • Availability
  • Add to Cart