ENA448968622 Enamine

4-(pyrazin-2-yl)-1,3-thiazol-2-amine

  • MDL number MFCD05722061
  • Cas Number : 19847-11-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207-208
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212396
Availability details for
Quantity
  • Availability
  • Add to Cart