ENA448968623 Enamine

4-(pyridin-3-yl)-4H-1,2,4-triazol-3-amine

  • MDL number MFCD20706279
  • Cas Number : 91447-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212397

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212397
Availability details for
Quantity
  • Availability
  • Add to Cart