ENA448968624 Enamine

tert-butyl N-{2-[(2-hydroxyphenyl)carbamoyl]ethyl}carbamate

  • MDL number MFCD17839385
  • Cas Number : 867353-53-5
  • Molecular Weight : 280.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-212405

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212405
Availability details for
Quantity
  • Availability
  • Add to Cart