ENA448968625 Enamine

6-fluoro-1-[tris(propan-2-yl)silyl]-1H-indole

  • MDL number MFCD26384468
  • Cas Number : 908600-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212410
Availability details for
Quantity
  • Availability
  • Add to Cart