ENA448968659 Enamine

3-methyl-1,2-dihydropyridine-2-thione

  • MDL number MFCD25541694
  • Cas Number : 18368-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-212466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212466
Availability details for
Quantity
  • Availability
  • Add to Cart