ENA448968670 Enamine

2-amino-3-(pyrrolidin-1-yl)propan-1-ol dihydrochloride

  • MDL number MFCD28505903
  • Cas Number : 1803612-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212797

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212797
Availability details for
Quantity
  • Availability
  • Add to Cart